Diethylnitrosoamine
Catalog # EHERC15603500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:102.14 g/mol
EINECS:200-226-1
UN:2810
ADR:6.1,III
CAS Number:55-18-5
Formula:C₄H₁₀N₂O
MDL Number:MFCD00013890