Order Entry
Export
ContactUsLinkComponent
Methyl heptadecanoate
Methyl heptadecanoate
Catalog # EHERC14122650
Supplier:  EHRENSTORFER
CAS Number:  
Methyl heptadecanoate
Catalog # EHERC14122650
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :284.48 g/mol
EINECS:217-055-3
CAS Number:1731-92-6
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00009001