4-Bromo-2-methoxybenzoic acid
Glass bottle for solids, 25 g
Catalog # COBBCA-4764-25G
Supplier: COMBI-BLOCKS
4-Bromo-2-methoxybenzoic acid
Catalog # COBBCA-4764-25G
Supplier: COMBI-BLOCKS
CAS Number: 72135-36-5
Jump to:
Product Details & Documents
- CAS Number:72135-36-5
- Formula:C₈H₇BrO₃
- MDL Number:MFCD07780665
- Molecular Weight:231.05 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



