2,6-Dichlorophenylhydrazinium chloride
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride
- CAS Number:50709-36-9
- Formula:C₆H₇Cl₃N₂
- MDL Number:MFCD00012930
- Molecular Weight:213.49 g/mol
- EINECS:256-730-7
3 Options Available Below
Product Details & Documents
Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride
- CAS Number:50709-36-9
- Formula:C₆H₇Cl₃N₂
- MDL Number:MFCD00012930
- Molecular Weight:213.49 g/mol
- EINECS:256-730-7
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



