Order Entry
Export
Orders LinkContactUsLinkComponent
2-Fluorophenylhydrazinium chloride

2-Fluorophenylhydrazinium chloride

Supplier:   Thermo Scientific
2-Fluorophenylhydrazinium chloride
Supplier:  Thermo Scientific
Certificates
2-Fluorophenylhydrazinium chloride

3 Options Available Below

Product Details & Documents
Synonyms:  (2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride
  • CAS Number:2924-15-4
  • Formula:C₆H₈ClFN₂
  • MDL Number:MFCD00012927
  • Molecular Weight:162.59 g/mol
  • EINECS:220-885-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g