
1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4526-07-2
:
Formula:C₁₀H₁₈Si₂
MDL Number:MFCD00009839
:194.42 g/mol
Melting Pt:110…113 °C
Density:0.752
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
:GHS02