Methyl m-anisate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5368-81-0
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00017193
:166.18 g/mol
Boiling Pt:238 °C
Density:1.15
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:226-359-5