Benzophenone imine 95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 95% CAS No.: 1013-88-3 Molecular Formula: C13H11N Formula Weight: 181.23 Refractive Index: 1.6180 Boiling Point: 151-153°/10mm MDL No.: MFCD00001760
Synonyms: Benzophenoneimine
- CAS Number:1013-88-3
- Formula:C₁₃H₁₁N
- MDL Number:MFCD00001760
- Molecular Weight:181.24 g/mol
- Boiling Pt:151…153 °C (10 mmHg)
- Density:1.08
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 1013-88-3 Molecular Formula: C13H11N Formula Weight: 181.23 Refractive Index: 1.6180 Boiling Point: 151-153°/10mm MDL No.: MFCD00001760
Synonyms: Benzophenoneimine
- CAS Number:1013-88-3
- Formula:C₁₃H₁₁N
- MDL Number:MFCD00001760
- Molecular Weight:181.24 g/mol
- Boiling Pt:151…153 °C (10 mmHg)
- Density:1.08
- Storage Temperature:Ambient



