Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4,6-dimethoxy-1,3,5-triazine
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥98%
:
Thermo Fisher Scientific
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥98%
:
Thermo Fisher Scientific
:
3140-73-6
:
:
C₅H₆ClN₃O₂
:
MFCD00075607
:
175.57 g/mol
:
99…100 °C (1 mmHg)
:
75…79 °C
:
Ambient
:
221-541-0
:
GHS07
msds_ALFAB24474
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years