1,3-Dibenzoyloxybenzene ≥98%
Catalog # A11983.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:318.33 g/mol
Melting Pt:116…119 °C
Flash Pt:93 °C (199 °F)
Storage Temperature:Ambient
EINECS:202-294-8
CAS Number:94-01-9
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00016576