
2,4-Difluorophenylhydrazinium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51523-79-6
:
Formula:C₆H₇ClF₂N₂
MDL Number:MFCD00013383
:180.58 g/mol
Melting Pt:∼ 255 °C (dec.)
:GHS07
Storage Temperature:Ambient