

3,5-Dimethylphenylhydrazinium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60481-36-9
:
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00052269
:172.66 g/mol
Melting Pt:180 °C (dec.)
:GHS06
:GHS07
Storage Temperature:Ambient