0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,3-Dichlorophenylhydrazinium chloride
2,3-Dichlorophenylhydrazinium chloride ≥97%
2,3-Dichlorophenylhydrazinium chloride ≥97%
:
Thermo Fisher Scientific
2,3-Dichlorophenylhydrazinium chloride ≥97%
:
Thermo Fisher Scientific
:
21938-47-6
:
:
C₆H₇Cl₃N₂
:
MFCD00052265
:
∼ 204 °C (dec.)
:
213.49 g/mol
:
GHS06
:
GHS07
:
Ambient
msds_ALFAA10717
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years