Order Entry
Export
ContactUsLinkComponent
1,3-Dibenzoylpropane 99%
1,3-Dibenzoylpropane 99%
Catalog # ACRO409010100
CAS Number:  
undefined
1,3-Dibenzoylpropane 99%
Catalog # ACRO409010100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 67 °C
  • GC:
    ≥98.5 %
  • Volatile matter:
    ≤4 % (110 °C)

Specification Test Results

About this item

 :
 :252.31 g/mol
Melting Pt:64…67 °C
Storage Temperature:Ambient
CAS Number:6263-83-8
Formula:C₁₇H₁₆O₂
MDL Number:MFCD00051401