Benzophenone imine 97% stabilised
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Benzophenoneimine
- CAS Number:1013-88-3
- Formula:C₁₃H₁₁N
- MDL Number:MFCD00001760
- Molecular Weight:181.24 g/mol
- Boiling Pt:216 °C (75 mmHg)
- Melting Pt:–30 °C
- Density:1.08 g/cm³
- Flash Pt:145 °C
- Storage Temperature:Ambient
- ADR:9,III
3 Options Available Below
Product Details & Documents
Synonyms: Benzophenoneimine
- CAS Number:1013-88-3
- Formula:C₁₃H₁₁N
- MDL Number:MFCD00001760
- Molecular Weight:181.24 g/mol
- Boiling Pt:216 °C (75 mmHg)
- Melting Pt:–30 °C
- Density:1.08 g/cm³
- Flash Pt:145 °C
- Storage Temperature:Ambient
- ADR:9,III
Documents
ACRO36859
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6170 to 1.6190 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:Approx. 0.02 % w/w Hydroquinone



