Order Entry
Export
Orders LinkContactUsLinkComponent
 

Benzophenone oxime 98%

Irritant
Corrosive
   Sustainable Options Available
Benzophenone oxime 98%
   Sustainable Options Available
Certificates
Benzophenone oxime 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:574-66-3
  • Formula:C₁₃H₁₁NO
  • MDL Number:MFCD00051461
  • Molecular Weight:197.24 g/mol
  • Melting Pt:140…144 °C
  • Storage Temperature:Freezer
  • EINECS:209-373-6

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    140 to 144 °C
  • HPLC:
    ≥97.5 %

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 30,30 €
    30,30 €
    1 * 5 g
       30,30 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    342330050.
      
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 93,20 €
    93,20 €
    1 * 25 g
       93,20 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    342330250.