4-(4-Nitrobenzyl)pyridine 98%
: 4-(p-Nitrobenzyl)Pyridine
- :1083-48-3
- :C₁₂H₁₀N₂O₂
- :MFCD00006445
- :214.22 g/mol
- :69…72 °C
- :Ambient
- :214-108-2
: 4-(p-Nitrobenzyl)Pyridine
- :1083-48-3
- :C₁₂H₁₀N₂O₂
- :MFCD00006445
- :214.22 g/mol
- :69…72 °C
- :Ambient
- :214-108-2
- Appearance:Light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:69 to 72 °C
- Titration with HClO4:≥97.5 %
Product Family Options
- Pack typePk



