trans-1,4-Diphenyl-2-butene-1,4-dione 97%
:959-28-4
:
:C₁₆H₁₂O₂
:MFCD00003083
:236.27 g/mol
:108…112 °C
:Ambient
:213-498-1
- Appearance:Yellow to light orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:108 to 112 °C
- GC:≥96.0 %