



1-Bromo-3,5-difluorobenzene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:461-96-1
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00010305
:192.99 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–27 °C
Density:1.676 g/cm³ (20 °C)
EINECS:416-710-2
UN:1993
ADR:3,III
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient