Order Entry
Export
ContactUsLinkComponent
4-Bromo-1,2-difluorobenzene
4-Bromo-1,2-difluorobenzene
Catalog # MOLEM15984111
Supplier:  Molekula
CAS Number:  
4-Bromo-1,2-difluorobenzene
Catalog # MOLEM15984111
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :192.99 g/mol
Boiling Pt:150 °C (1013 hPa)
Melting Pt:–4 °C
Density:1.7 g/cm³ (20 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
EINECS:206-481-5
UN:1993
ADR:3,III
CAS Number:348-61-8
Formula:C₆H₃BrF₂
MDL Number:MFCD00000304