
2-Bromo-1,4-difluorobenzene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:399-94-0
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00000345
:192.99 g/mol
Boiling Pt:158 °C (1013 hPa)
Melting Pt:–32 °C
Density:1,7414 g/cm³ (20 °C)
EINECS:206-920-0
:GHS07