Order Entry
Export
ContactUsLinkComponent
1,2-Dibromotetrafluorobenzene 99%
Irritant
1,2-Dibromotetrafluorobenzene 99%
Certificates
undefined
Irritant
1,2-Dibromotetrafluorobenzene 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5145 to 1.5175 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information