0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,2-Dibromo-4-fluorobenzene
1,2-Dibromo-4-fluorobenzene ≥98%
1,2-Dibromo-4-fluorobenzene ≥98%
1,2-Dibromo-4-fluorobenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1,2-Dibromo-4-fluorobenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
2369-37-1
:
Formula:
C₆H₃Br₂F
MDL Number:
MFCD00070780
:
253.9 g/mol
Storage Temperature:
Ambient
EINECS:
219-131-1
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20935
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation