
1-Bromo-3,5-dichlorobenzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19752-55-7
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000584
:225.9 g/mol
Boiling Pt:232 °C
Melting Pt:73…77 °C
Storage Temperature:Ambient
EINECS:243-270-7
:GHS07