
4-Bromo-1,2-dichlorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18282-59-2
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00040849
:225.9 g/mol
Boiling Pt:140…142 °C (30 mmHg)
Melting Pt:22…25 °C
Density:1.781
Flash Pt:87 °C (188 °F)
EINECS:242-160-6
:GHS07
Storage Temperature:Ambient