Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-3,5-dihydroxybenzoic acid
4-Bromo-3,5-dihydroxybenzoic acid
4-Bromo-3,5-dihydroxybenzoic acid
MOLEM16601852
:
Molekula
:
4-Bromo-3,5-dihydroxybenzoic acid
MOLEM16601852
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
233.02 g/mol
:
240-606-4
:
GHS07
:
16534-12-6
:
C₇H₅BrO₄
:
MFCD00002513