
5-Bromo-2,4-dihydroxybenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7355-22-8
:
Formula:C₇H₅BrO₄
MDL Number:MFCD00002452
:233.02 g/mol
Melting Pt:∼205 °C (dec.)
EINECS:230-881-9
:GHS07
Storage Temperature:Ambient