Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromobenzoic acid

4-Bromobenzoic acid

Glass bottle for solids, 100 g
Catalog # MOLEM82916461
Irritant
Supplier:   Molekula
4-Bromobenzoic acid
Catalog # MOLEM82916461
Supplier:  Molekula
CAS Number:  586-76-5
4-Bromobenzoic acid

Glass bottle for solids, 100 g

CAS Number:  586-76-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Bromobenzoic Acid
  • CAS Number:586-76-5
  • Formula:C₇H₅BrO₂
  • MDL Number:MFCD00002529
  • Molecular Weight:201.02 g/mol
  • Merck Index:13,01392
  • Boiling Pt:300 °C (1013 hPa)
  • Melting Pt:252…256 °C
  • Density:1.894 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-581-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications