4-Bromobenzonitrile
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: p-Bromobenzonitrile
- CAS Number:623-00-7
- Formula:C₇H₄BrN
- MDL Number:MFCD00001811
- Molecular Weight:182.02 g/mol
- Boiling Pt:236 °C (1013 hPa)
- Melting Pt:∼90 °C
- Density:1.562 g/cm³ (20 °C)
- Flash Pt:130 °C
- Storage Temperature:Ambient
- EINECS:210-764-9
- UN:3439
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: p-Bromobenzonitrile
- CAS Number:623-00-7
- Formula:C₇H₄BrN
- MDL Number:MFCD00001811
- Molecular Weight:182.02 g/mol
- Boiling Pt:236 °C (1013 hPa)
- Melting Pt:∼90 °C
- Density:1.562 g/cm³ (20 °C)
- Flash Pt:130 °C
- Storage Temperature:Ambient
- EINECS:210-764-9
- UN:3439
- ADR:6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



