
3,5-Dibromo-4-hydroxybenzaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2973-77-5
:
Formula:C₇H₄Br₂O₂
MDL Number:MFCD00016980
:279.92 g/mol
Melting Pt:179…182 °C
EINECS:221-017-1
:GHS07
Storage Temperature:Ambient