Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,5-Dibromosalicylaldehyde

Glass bottle for solids, 25 g
Catalog # MOLEM32225469
Irritant
Supplier:   Molekula
3,5-Dibromosalicylaldehyde
Catalog # MOLEM32225469
Supplier:  Molekula
CAS Number:  90-59-5
3,5-Dibromosalicylaldehyde

Glass bottle for solids, 25 g

CAS Number:  90-59-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,5-Dibromo-2-hydroxybenzaldehyde
  • CAS Number:90-59-5
  • Formula:C₇H₄Br₂O₂
  • MDL Number:MFCD00003318
  • Molecular Weight:279.92 g/mol
  • Merck Index:13,03052
  • Boiling Pt:261 °C (1013 hPa)
  • Melting Pt:86 °C
  • EINECS:202-003-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications