Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dibromosalicylaldehyde ≥98%

3,5-Dibromosalicylaldehyde ≥98%

3,5-Dibromosalicylaldehyde ≥98%
Certificates
3,5-Dibromosalicylaldehyde ≥98%
3,5-Dibromosalicylaldehyde ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 90-59-5 Molecular Formula: C7H4Br2O2 Formula Weight: 279.93 Melting Point: 81-84° Beilstein Reference: 1424739 Merck Reference: 14,3027 MDL No.: MFCD00003318

Synonyms:  3,5-Dibromo-2-hydroxybenzaldehyde
  • CAS Number:90-59-5
  • Formula:C₇H₄Br₂O₂
  • MDL Number:MFCD00003318
  • Molecular Weight:279.92 g/mol
  • Merck Index:14,3027
  • Boiling Pt:261 °C (1013 hPa)
  • Melting Pt:81…84 °C
  • Storage Temperature:Ambient
  • EINECS:202-003-4
Product Family Options

Product Information