Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Earplugs
Banded Earplugs
Replacement plugs for banded earplugs, E-A-R™ Swerve™
Replacement plugs for banded earplugs, E-A-R™ Swerve™
Replacement plugs for banded earplugs, E-A-R™ Swerve™
Catalog # MMMASW01001
Supplier:
3M
Expand image
Replacement plugs for banded earplugs, E-A-R™ Swerve™
Catalog # MMMASW01001
Supplier:
3M
Specifications
Colour:
Yellow
Description:
Replacement pods for Earplugs E-A-R™ Swerve™
Pk:
100 PAIR
Specifications
About this item
Product Details
Product Details