Order Entry
Export
ContactUsLinkComponent
1,3-Dibromobenzene
Irritant
1,3-Dibromobenzene
Catalog # EHERC12216711
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
1,3-Dibromobenzene
Catalog # EHERC12216711
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :235.91 g/mol
Boiling Pt:218 °C (1013 hPa)
Melting Pt:–7 °C
Density:1,952 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-574-2
 :GHS07
CAS Number:108-36-1
Formula:C₆H₄Br₂
MDL Number:MFCD00000078