Order Entry
Export
Orders LinkContactUsLinkComponent
9,10-Diphenylanthracene

9,10-Diphenylanthracene

Glass bottle for solids, 5 g
Catalog # MOLE23026231-5G
Supplier:   Molekula
9,10-Diphenylanthracene
Catalog # MOLE23026231-5G
Supplier:  Molekula
CAS Number:  1499-10-1
9,10-Diphenylanthracene

Glass bottle for solids, 5 g

CAS Number:  1499-10-1
Price
180,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1499-10-1
  • Formula:C₂₆H₁₈
  • MDL Number:MFCD00001253
  • Molecular Weight:330.43 g/mol
  • Boiling Pt:485 °C (1013 hPa)
  • Melting Pt:245 °C
  • Storage Temperature:Ambient
  • EINECS:216-105-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications