Order Entry
Export
ContactUsLinkComponent
9-Anthrealdehyde
9-Anthrealdehyde
Catalog # MOLE30708426-100G
Supplier:  Molekula
CAS Number:  
9-Anthrealdehyde
Catalog # MOLE30708426-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde
 :
 :206.24 g/mol
Boiling Pt:369 °C (1013 hPa)
Melting Pt:101…103 °C
Storage Temperature:Ambient
EINECS:211-383-0
CAS Number:642-31-9
Formula:C₁₅H₁₀O
MDL Number:MFCD00001254