3,7-Dimethyl-1-octene, Sigma-Aldrich®
Catalog # SIAL40980-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:140.27 g/mol
Boiling Pt:154…156 °C (1013 hPa)
Melting Pt:–103 °C
Density:0.733 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:225-635-2
UN:3295
ADR:3,III
CAS Number:4984-01-4
Formula:C₁₀H₂₀