Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Chloro-2-methylpropene

Glass bottle for solids, 25 g
Catalog # MOLE11829435-25G
Flammable
Corrosive
Irritant
Environment
Supplier:   Molekula
3-Chloro-2-methylpropene
Catalog # MOLE11829435-25G
Supplier:  Molekula
CAS Number:  563-47-3
3-Chloro-2-methylpropene

Glass bottle for solids, 25 g

CAS Number:  563-47-3
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:563-47-3
  • Formula:C₄H₇Cl
  • MDL Number:MFCD00000953
  • Molecular Weight:90.55 g/mol
  • Merck Index:13,02167
  • Boiling Pt:72 °C (1013 hPa)
  • Melting Pt:–80 °C
  • Density:0.93 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-251-2
  • UN:2554
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications