1-Tridecene ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 2437-56-1 Molecular Formula: C13H26 Formula Weight: 182.35 Flash Point: 79°(174°F) Refractive Index: 1.4330 Odour: Mild Melting Point: -23° Boiling Point: 232-233° Beilstein Reference: 1744660 MDL No.: MFCD00008976
- CAS Number:2437-56-1
- Formula:C₁₃H₂₆
- MDL Number:MFCD00008976
- Molecular Weight:182.35 g/mol
- Boiling Pt:232…233 °C
- Melting Pt:–23 °C
- Density:0.766
- Flash Pt:79 °C (174 °F)
- Storage Temperature:Ambient
- EINECS:219-443-8
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 2437-56-1 Molecular Formula: C13H26 Formula Weight: 182.35 Flash Point: 79°(174°F) Refractive Index: 1.4330 Odour: Mild Melting Point: -23° Boiling Point: 232-233° Beilstein Reference: 1744660 MDL No.: MFCD00008976
- CAS Number:2437-56-1
- Formula:C₁₃H₂₆
- MDL Number:MFCD00008976
- Molecular Weight:182.35 g/mol
- Boiling Pt:232…233 °C
- Melting Pt:–23 °C
- Density:0.766
- Flash Pt:79 °C (174 °F)
- Storage Temperature:Ambient
- EINECS:219-443-8



