trans-1,4-Dichloro-2-butene
Catalog # MOLEM51671550
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:125 g/mol
Boiling Pt:156 °C (1013 hPa)
Melting Pt:1 °C
Density:1.1873 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:203-779-7
UN:3390
ADR:6.1,I
CAS Number:110-57-6
Formula:C₄H₆Cl₂