Prefiltration capsules, Sartopure® GF Plus
Supplier: Sartorius
Certificates
About this item
Glass fibre, PP support layers. They are self-contained, ready to use filter units for removal of contaminants like colloids, lipids, protein aggregates and particles from bio-pharmaceutical fluids. They are used for protection of membrane filters, chromatography- and ultrafiltration systems.
- Feature with highly charged glass fiber layers for effective clarification of fluid streams
- Adsorptive depth filters assures outstanding total throughput performance
Specifications
- Catalog No:
- SARP5555303P8-SS-A
- SARP5555305P0-SS-V
- Type:
- Sartopure GF Size 8
- Sartopure GF Size 0
- Inlet/outlet:
- SS (1 1/2 inch Sanitary flange / 1 1/2 inch Sanitary flange)
- SS (1 1/2 inch Sanitary flange / 1 1/2 inch Sanitary flange)
- Ø×L:
- 77×197 mm
- 77×197 mm
- Max. pressure:
- 5 bar @ 20 °C
- 5 bar @ 20 °C