Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloropropene
2,3-Dichloropropene ≥98%
2,3-Dichloropropene ≥98%
:
Thermo Fisher Scientific
2,3-Dichloropropene ≥98%
:
Thermo Fisher Scientific
:
78-88-6
:
:
C₃H₄Cl₂
:
MFCD00000943
:
110.97 g/mol
:
93…96 °C
:
–83 °C
:
1.209
:
10 °C (50 °F)
:
201-153-8
:
2047
:
3,II
:
GHS02
:
GHS05
:
GHS08
:
GHS07
:
Ambient
msds_ALFAL08943
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years