



2,3-Dichloropropene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-88-6
:
Formula:C₃H₄Cl₂
MDL Number:MFCD00000943
:110.97 g/mol
Boiling Pt:94 °C (760 mmHg)
Melting Pt:–83.1 °C
Density:1.2 g/cm³
Flash Pt:10 °C
EINECS:201-153-8
UN:2047
ADR:3,II
:GHS05
:GHS08
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to pink-brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4601 to 1.4621 (20°C, 589 nm)
- GC:≥97.5 %
- UV:(3 % in water)
- Chloride (Cl):≤500 ppm