Order Entry
Export
Orders LinkContactUsLinkComponent
3,3-Dimethylpentane 99%

3,3-Dimethylpentane 99%

Irritant
Health
Environment
Flammable
3,3-Dimethylpentane 99%
Certificates
3,3-Dimethylpentane 99%

3 Options Available Below

Product Details & Documents
  • CAS Number:562-49-2
  • Formula:C₇H₁₆
  • MDL Number:MFCD00009322
  • Molecular Weight:100.2 g/mol
  • Boiling Pt:86 °C (760 mmHg)
  • Melting Pt:–135 °C
  • Density:0.69 g/cm³
  • Flash Pt:–6 °C
  • Storage Temperature:Ambient
  • EINECS:209-230-8
  • ADR:3,II
  •  :Density: 0.6936 g/cm³ (20 °C)
  • UN:1206

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3900 to 1.3920 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 45,40 €
    45,40 €
    1 * 1 g
       45,40 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 149,00 €
    149,00 €
    1 * 5 g
       149,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 570,00 €
    570,00 €
    1 * 25 g
       570,00 €