

6-Methyl-2,4-heptanedione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3002-23-1
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00008939
:142.2 g/mol
Boiling Pt:183 °C
Density:0.919
Flash Pt:58 °C (136 °F)
Storage Temperature:Ambient
EINECS:221-093-6
UN:1224
ADR:3,III
:GHS02
:GHS07