Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Methyl-2-pentanone

Flammable
Irritant
Supplier:   Roth Carl
4-Methyl-2-pentanone
Supplier:  Roth Carl
Certificates
4-Methyl-2-pentanone
4-Methyl-2-pentanone

2 Options Available Below

Product Details & Documents
Synonyms:  Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
  • CAS Number:108-10-1
  • Formula:(CH₃)₂CHCH₂COCH₃
  • MDL Number:MFCD00008938
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,05227
  • Boiling Pt:117.4 °C (1013 hPa)
  • Melting Pt:–84 °C
  • Density:0.801 g/cm³ (20 °C)
  • Flash Pt:16 °C
  • Storage Temperature:Ambient
  • EINECS:203-550-1
  • UN:1245
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 134,00 €
    134,00 €
    1 * 1 L
       134,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L