Order Entry
Export
Orders LinkContactUsLinkComponent
 

n-Undecane

Flammable
Health
Supplier:   Roth Carl
n-Undecane
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Undecane
  • CAS Number:1120-21-4
  • Formula:H₃C(CH₂)₉CH₃
  • MDL Number:MFCD00008959
  • Molecular Weight:156.31 g/mol
  • Boiling Pt:196 °C (1013 hPa)
  • Melting Pt:–26.7 °C
  • Density:0.7416 g/cm³ (20 °C)
  • Flash Pt:60 °C
  • Storage Temperature:Ambient
  • EINECS:214-300-6
  • UN:2330
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL