Order Entry
Export
Orders LinkContactUsLinkComponent
n-Undecane 99%

n-Undecane 99%

n-Undecane 99%
Certificates
n-Undecane 99%

4 Options Available Below

Product Details & Documents
Synonyms:  Undecane
  • CAS Number:1120-21-4
  • Formula:H₃C(CH₂)₉CH₃
  • MDL Number:MFCD00008959
  • Molecular Weight:156.31 g/mol
  • Boiling Pt:196 °C (760 mmHg)
  • Melting Pt:–26 °C
  • Density:0.74 g/cm³
  • Flash Pt:65 °C
  • Storage Temperature:Ambient
  • EINECS:214-300-6
  • UN:2330
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4160 to 1.4180 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information