n-Tridecane
Catalog # PROOCHE153
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
:184.37 g/mol
Boiling Pt:235.4 °C (1013 hPa)
Melting Pt:–5.5 °C
Density:0.7564 g/cm³ (20 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:211-093-4
UN:3295
ADR:3,III
:GHS08
CAS Number:629-50-5
Formula:H₃C(CH₂)₁₁CH₃
MDL Number:MFCD00008979