
n-Tridecane for synthesis, Sigma-Aldrich®
8.14631.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
- Solubility:(25 °C) practically insoluble
- Melting Point:-5 °C
- Molar Mass:184.36 g/mol
- Boiling Point:235 °C (1013 hPa)
- Vapor Pressure:- 0.5 hPa (20 °C)
- Flash Point:79 °C
- Explosion Limit:0.6 - 6.5 %(V)
- Density:0.757 g/cm3 (20 °C)
- Ignition Point:204 °C
:
:184.37 g/mol
:235.4 °C (1013 hPa)
:–5.5 °C
:0.7564 g/cm³ (20 °C)
:79 °C
:Ambient
:211-093-4
:3295
:3,III
:M-Clarity™ quality level = MQ200
:GHS08
:629-50-5
:H₃C(CH₂)₁₁CH₃
:MFCD00008979